missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Acetyl N-Benzyloxycarbonyl Valganciclovir, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
O-Acetyl N-Benzyloxycarbonyl Valganciclovir
Specifications
Specifications
| Chemical Name or Material | O-Acetyl N-Benzyloxycarbonyl Valganciclovir |
| CAS | 194159-22-3 |
| Analyte or Component Names | O-Acetyl N-Benzyloxycarbonyl Valganciclovir |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H30 N6 O8 |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Valine, N-[(phenylmethoxy)carbonyl]-, 3-(acetyloxy)-2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]propyl ester, [3-acetoxy-2-[(2-amino-6-oxo-3H-purin-9-yl)methoxy]propyl] (2S)-2-(benzyloxycarbonylamino)-3-methyl-butanoate, O-Acetyl N-CBZ Valganciclovir, O-Acetyl N-Benzyloxycarbonyl Valganciclovir |
| InChI Formula | InChI=1S/C24H30N6O8/c1-14(2)18(27-24(34)37-9-16-7-5-4-6-8-16)22(33)36-11-17(10-35-15(3)31)38-13-30-12-26-19-20(30)28-23(25)29-21(19)32/h4-8,12,14,17-18H,9-11,13H2,1-3H3,(H,27,34)(H3,25,28,29,32)/t17?,18-/m0/s1 |
| SMILES | CC(C)[C@H](NC(=O)OCc1ccccc1)C(=O)OCC(COC(=O)C)OCn2cnc3C(=O)N=C(N)Nc23 |
| IUPAC Name | [3-acetyloxy-2-[(2-amino-6-oxo-3H-purin-9-yl)methoxy]propyl] (2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction