missing translation for 'onlineSavingsMsg'
Learn More

O-Acetyl Losartan, TRC

Product Code. 30316003
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316003 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316003 Supplier Toronto Research Chemicals Supplier No. TRCA185300250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O-Acetyl Losartan
CAS 1006062-27-6
Molecular Formula C24 H25 Cl N6 O2
Quantity 250 mg
Synonym [2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-5-yl]methyl Acetate, Losartan Potassium Imp. J (EP)
InChI Formula InChI=1S/C24H25ClN6O2/c1-3-4-9-22-26-23(25)21(15-33-16(2)32)31(22)14-17-10-12-18(13-11-17)19-7-5-6-8-20(19)24-27-29-30-28-24/h5-8,10-13H,3-4,9,14-15H2,1-2H3,(H,27,28,29,30)
SMILES CCCCc1nc(Cl)c(COC(=O)C)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4
IUPAC Name [2-butyl-5-chloro-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methyl acetate
Molecular Weight (g/mol) 464.95
Formula Weight 464.1728
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.