missing translation for 'onlineSavingsMsg'
Learn More

O-(4-Methoxybenzyl)hydroxylamine hydrochloride, 98%

Product Code. 15458236
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458236 1 g 1g
15468236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458236 Supplier Thermo Scientific Alfa Aesar Supplier No. H63312.03
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 876-33-5 | C8H12ClNO2 | 189.64 g/mol

O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 876-33-5
MDL Number MFCD01114582
Synonym o-4-methoxybenzyl hydroxylamine, o-4-methoxyphenyl methyl hydroxylamine, p-methoxybenzyloxyamine, hydroxylamine, o-p-methoxybenzyl, hydroxylamine, o-4-methoxyphenyl methyl, o-p-methoxybenzyl hydroxylamine, o-p-methoxybenzyl-hydroxylamine, o-4-methoxy-benzyl-hydroxylamine, 1-aminooxy methyl-4-methoxybenzene, 1-aminooxy methyl 4-methoxy-benzene
PubChem CID 120426
IUPAC Name O-[(4-methoxyphenyl)methyl]hydroxylamine

Specifications

Color White
Molecular Formula C8H11NO2
Quantity 1 g
InChI Key MVSMBIBGGPSEHQ-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)CON
Molecular Weight (g/mol) 153.181
Formula Weight 153.18
Percent Purity 98%
Physical Form Solid
Chemical Name or Material O-(4-Methoxybenzyl)hydroxylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.