missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-3M3FBS, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Inactive analog of m-3M3FBS. Used as a negative control.
Active Analog also available.
Specifications
Specifications
| CAS | 313981-55-4 |
| Molecular Formula | C16H16F3NO2S |
| MDL Number | MFCD00784362 |
| Quantity | 10 mg |
| Synonym | o-3m3fbs, 2,4,6-trimethyl-n-2-trifluoromethyl phenyl benzenesulfonamide, tocris-1942, benzenesulfonamide,2,4,6-trimethyl-n-2-trifluoromethyl phenyl |
| InChI Key | SKJJIFRWCCSXGL-UHFFFAOYSA-N |
| SMILES | CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC=C1C(F)(F)F |
| IUPAC Name | 2,4,6-trimethyl-N-[2-(trifluoromethyl)phenyl]benzene-1-sulfonamide |
| Molecular Weight (g/mol) | 343.36 |
| PubChem CID | 770820 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction