missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-304, MedChemExpress
Description
O-304 is a first-in-class, orally available pan-AMPK activator, which increases AMPK activity by suppressing the dephosphorylation of pAMPK. O-304 exhibits a great potential as a drug to treat type 2 diabetes (T2D) and associated cardiovascular complications .
Specifications
Specifications
| Chemical Name or Material | O-304 |
| Physical Form | Solid |
| CAS | 1261289-04-6 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C16H11Cl2N3O2S |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : 30 mg/mL (78.90 mM; Need ultrasonic and warming) |
| SMILES | O=C(NC(SN1CC2=CC=C(Cl)C=C2)=NC1=O)C3=CC=C(Cl)C=C3 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?