missing translation for 'onlineSavingsMsg'
Learn More

O-304, MedChemExpress

Product Code. 16415456
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16415456 100 mg 100mg
16487317 5 mg 5mg
16463156 25 mg 25mg
16434608 10 mg 10mg
16430907 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16415456 Supplier Medchem Express Supplier No. HY112233

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress O-304 is a first-in-class, orally available pan-AMPK activator, which increases AMPK activity by suppressing the dephosphorylation of pAMPK. O-304 exhibits a great potential as a drug to treat type 2 diabetes (T2D) and associated cardiovascular complications .

O-304 is a first-in-class, orally available pan-AMPK activator, which increases AMPK activity by suppressing the dephosphorylation of pAMPK. O-304 exhibits a great potential as a drug to treat type 2 diabetes (T2D) and associated cardiovascular complications .

Specifications

Chemical Name or Material O-304
Physical Form Solid
CAS 1261289-04-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C16H11Cl2N3O2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 30 mg/mL (78.90 mM; Need ultrasonic and warming)
SMILES O=C(NC(SN1CC2=CC=C(Cl)C=C2)=NC1=O)C3=CC=C(Cl)C=C3
Molecular Weight (g/mol) 380.25
Formula Weight 380.25
Percent Purity 99.35%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.