Learn More
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 99+%, Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, >-99%, C7H5ClF5NO, CAS Number-57981-02-9
£28.10 - £276.00
Chemical Identifiers
CAS | 57981-02-9 |
---|---|
Molecular Formula | C7H5ClF5NO |
Molecular Weight (g/mol) | 249.57 |
MDL Number | MFCD00012953 |
InChI Key | HVMVKNXIMUCYJA-UHFFFAOYSA-N |
Synonym | o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv |
PubChem CID | 122307 |
IUPAC Name | O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride |
SMILES | [H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11488287
|
Thermo Scientific Alfa Aesar
A18368.01 |
0.25 g |
£28.10
0.25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468287
|
Thermo Scientific Alfa Aesar
|
1 g |
£76.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478287
|
Thermo Scientific Alfa Aesar
A18368.06 |
5 g |
£276.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride is used in the preparation of oximes of steroids having keto group. It is used as a derivatization reagent in the determination of thromboxane B2, prostaglandins, amygdalin and a variety of aldehydes, ketones and acids. It is a versatile reagent utilized for the detection of carbonyl-containing compounds by gas chromatography (GC), gas chromatography mass spectrometry (GC-MS) and other methods. As a derivatization reagent, it is also used for EC/LC analysis of keto-steroids and carbohydrates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsO-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride is used in the preparation of oximes of steroids having keto group. It is used as a derivatization reagent in the determination of thromboxane B2, prostaglandins, amygdalin and a variety of aldehydes, ketones and acids. It is a versatile reagent utilized for the detection of carbonyl-containing compounds by gas chromatography (GC), gas chromatography mass spectrometry (GC-MS) and other methods. As a derivatization reagent, it is also used for EC/LC analysis of keto-steroids and carbohydrates.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
57981-02-9 | |
249.57 | |
HVMVKNXIMUCYJA-UHFFFAOYSA-N | |
122307 | |
[H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F |
C7H5ClF5NO | |
MFCD00012953 | |
o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv | |
O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride |
Specifications
57981-02-9 | |
C7H5ClF5NO | |
4031190 | |
Soluble in water. | |
[H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F | |
249.57 | |
249.57 | |
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride |
∼215°C (sublimation) | |
MFCD00012953 | |
o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv | |
HVMVKNXIMUCYJA-UHFFFAOYSA-N | |
O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride | |
122307 | |
≥99% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 261-057-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only