missing translation for 'onlineSavingsMsg'
Learn More

O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 99%

Product Code. 11303097
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11303097

Brand: Thermo Scientific Alfa Aesar L13470.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 125700-67-6 | C11H16BF4N5O | 321.09 g/mol

O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate is used as used for solid-phase peptide synthesis. It is also used to study ten different phosphonium and uronium salt-based coupling reagents, which are used to synthesize four peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(1H-Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate is used as used for solid-phase peptide synthesis. It is also used to study ten different phosphonium and uronium salt-based coupling reagents, which are used to synthesize four peptides.

Solubility
Soluble in acetonitrile (100 mg/ml, clear, colorless), DMF (160.55 mg/ml, clear), and water (3 mg/ml at 20°C).

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2-8°C. Sensitive to light and moisture. Store under inert gas.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 125700-67-6
Molecular Formula C11H16BF4N5O
Molecular Weight (g/mol) 321.09
MDL Number MFCD00077413
InChI Key JKEKMBGUVUKMQB-UHFFFAOYSA-N
Synonym tbtu, 2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, 2-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-1h-benzotriazol-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-benzotriazol-1-yl uronium tetrafluoroborate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluroniumtetrafluoroborate, o-benzotriazol-1-yl-n,n,n',n'-tetra methyluronium tetrafluoroborate
PubChem CID 2733207
SMILES F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C12)=[N+](C)C

Specifikationer

Melting Point 195°C to 198°C (decomposition)
Quantity 5 g
Beilstein 7066325
Solubility Information Soluble in acetonitrile (100mg/ml,clear,colorless),DMF (160.55mg/ml,clear),and water (3mg/ml at 20°C).
IUPAC Name [(1H-1,2,3-benzotriazol-1-yloxy)(dimethylamino)methylidene]dimethylazanium; tetrafluoroboranuide
Formula Weight 321.08
Percent Purity 99%
Chemical Name or Material O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.