missing translation for 'onlineSavingsMsg'
Learn More

O-(1-(4-Fluorophenyl)butan-1-one) Haloperidol, TRC

Product Code. 30396838
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396838 10 mg 10mg
30398092 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396838 Supplier Toronto Research Chemicals Supplier No. TRCF59622510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O-(1-(4-Fluorophenyl)butan-1-one) Haloperidol
Analyte or Component Names O-(1-(4-Fluorophenyl)butan-1-one) Haloperidol
Shipping Condition Room Temperature
Molecular Formula C31 H32 Cl F2 N O3
Synonym 4-((4-(4-Chlorophenyl)-1-(4-(4-fluorophenyl)-4-oxobutyl)piperidin-4-yl)oxy)-1-(4-fluorophenyl)butan-1-one
InChI Formula InChI=1S/C31H32ClF2NO3/c32-26-11-9-25(10-12-26)31(38-22-2-4-30(37)24-7-15-28(34)16-8-24)17-20-35(21-18-31)19-1-3-29(36)23-5-13-27(33)14-6-23/h5-16H,1-4,17-22H2
Solution Type Neat
SMILES Fc1ccc(cc1)C(=O)CCCOC2(CCN(CCCC(=O)c3ccc(F)cc3)CC2)c4ccc(Cl)cc4
IUPAC Name 4-[4-(4-chlorophenyl)-4-[4-(4-fluorophenyl)-4-oxobutoxy]piperidin-1-yl]-1-(4-fluorophenyl)butan-1-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 540.04
Quantity 10 mg
Formula Weight 539.204
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.