missing translation for 'onlineSavingsMsg'
Learn More

Nystose, MedChemExpress

Product Code. 16494917
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494917 50 mg 50mg
16449166 100 mg 100mg
16419238 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494917 Supplier Medchem Express Supplier No. HYN1499
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nystose is a tetrasaccharide with two fructose molecules linked via beta (1→2) bonds to the fructosyl moiety of sucrose.

Nystose is a tetrasaccharide with two fructose molecules linked via beta (1→2) bonds to the fructosyl moiety of sucrose.

Specifications

Chemical Name or Material Nystose
CAS 13133-07-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H42O21
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (150.02 mM; Need ultrasonic) ∣H2O : 83.33 mg/mL (125.01 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@](CO[C@@]2(O[C@H](CO)[C@@H](O)[C@@H]2O)CO[C@@]3(O[C@H](CO)[C@@H](O)[C@@H]3O)CO)(O[C@H](CO)[C@H]1O)O[C@H]([C@@H]([C@@H](O)[C@@H]4O)O)O[C@@H]4CO
Molecular Weight (g/mol) 666.58
Formula Weight 666.58
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.