missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nystatin, MedChemExpress
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
Description
Nystatin is an orally active polyene antifungal antibiotic effective against yeast and mycoplasma. Nystatin increases the permeability of plasma membranes to small monovalent ions, including chloridion. Nystatin is a cholesterol-sequestering agent, partially prevents Oxaliplatin-induced lipid raft aggregation, DR4 and DR5 clustering, and thereby reduces apoptosis.
Specifications
Specifications
| Chemical Name or Material | Nystatin |
| Physical Form | Solid |
| CAS | 1400-61-9 |
| Color | Light Yellow |
| Quantity | 200 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C47H75NO17 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : 50 mg/mL (53.99 mM; Need ultrasonic) |
| SMILES | C[C@@H]1OC(C[C@H](O)C[C@H](O)C[C@H](O)CC[C@@H](O)[C@H](O)C[C@@](O)(O[C@H]2C[C@H](/C=C/C=C/C=C/C=C/CC/C=C/C=C/[C@@H]([C@@H](O)[C@H]1C)C)O[C@H]3[C@@H](O)[C@@H](N)[C@H](O)[C@@H](C)O3)C[C@H](O)[C@H]2C(O)=O)=O.[Nystatin A1] |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction