missing translation for 'onlineSavingsMsg'
Learn More

Nystatin, MedChemExpress

Product Code. 16411856
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16411856 500 mg 500mg
16451256 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16411856 Supplier Medchem Express Supplier No. HY17409
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nystatin is an orally active polyene antifungal antibiotic effective against yeast and mycoplasma. Nystatin increases the permeability of plasma membranes to small monovalent ions, including chloridion. Nystatin is a cholesterol-sequestering agent, partially prevents Oxaliplatin-induced lipid raft aggregation, DR4 and DR5 clustering, and thereby reduces apoptosis.

Nystatin is an orally active polyene antifungal antibiotic effective against yeast and mycoplasma. Nystatin increases the permeability of plasma membranes to small monovalent ions, including chloridion. Nystatin is a cholesterol-sequestering agent, partially prevents Oxaliplatin-induced lipid raft aggregation, DR4 and DR5 clustering, and thereby reduces apoptosis.

Specifications

Chemical Name or Material Nystatin
Physical Form Solid
CAS 1400-61-9
Color Light Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C47H75NO17
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 50 mg/mL (53.99 mM; Need ultrasonic)
SMILES C[C@@H]1OC(C[C@H](O)C[C@H](O)C[C@H](O)CC[C@@H](O)[C@H](O)C[C@@](O)(O[C@H]2C[C@H](/C=C/C=C/C=C/C=C/CC/C=C/C=C/[C@@H]([C@@H](O)[C@H]1C)C)O[C@H]3[C@@H](O)[C@@H](N)[C@H](O)[C@@H](C)O3)C[C@H](O)[C@H]2C(O)=O)=O.[Nystatin A1]
Molecular Weight (g/mol) 926.09
Formula Weight 926.09
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.