missing translation for 'onlineSavingsMsg'
Learn More

Nystatin, Fisher BioReagents

Product Code. 10034587
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10034587 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10034587 Supplier Fisher BioReagents Supplier No. BP29495
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alters membrane permeability and allows potassium ion leakage

TRUSTED_SUSTAINABILITY

Specifications

Type Nystatin
Physical Form Solid
CAS 1400-61-9
Source Streptomyces noursei
Loss on Drying < 5.0%
Packaging Amber Glass
Potency > 4400 U/mg
Quantity 5 g
Molecular Formula C47H75NO17
Merck Index 15, 6825
InChI Key VQOXZBDYSJBXMA-AWCIUYGFSA-N
SMILES CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O
IUPAC Name (1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carb
Molecular Weight (g/mol) 926.107
PubChem CID 133640190
Formula Weight 926.1
Chemical Name or Material Nystatin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.