missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nystatin, 85+%

Product Code. 15474871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474871 25 g 25g
15565156 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474871 Supplier Thermo Scientific Chemicals Supplier No. 455500250
  • ONLINE EXCLUSIVE PRICE
    £112.00
    £91.65 / 25g
    Save £20.35
    18% Off
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1400-61-9 | C47H75NO17 | 926.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1400-61-9
Molecular Formula C47H75NO17
Molecular Weight (g/mol) 926.107
InChI Key VQOXZBDYSJBXMA-AWCIUYGFSA-N
PubChem CID 133640190
IUPAC Name (1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carb
SMILES CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O

Specifications

Type Nystatin
Melting Point >160.0°C
Infrared Spectrum Authentic
Water 6% max.
Quantity 25 g
Solubility Information Solubility in water: practically insoluble. Other solubilities: freely soluble in dmf and dimethylsulfoxide, slightly soluble in methanol, practically insoluble in alcohol
Formula Weight 926.1
Percent Purity ≥85%
Chemical Name or Material Nystatin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.