Learn More
Masterflex™ Straight Fitting, Threaded Nipple, NPT(M) x NPT(M), Nylon
Product Code. 11761104
Click to view available options
Thread Size:
3/8 in. NPT
Unit Size:
Pack of 6
Product Code. 11761104
missing translation for 'mfr': Masterflex™ 3062329
Only null left
Add to basket
Add to basket
Cole-Parmer™ Straight Fitting, Threaded Nipple, NPT(M) x NPT(M), Nylon features good chemical resistance.
  • Easy to use
  • Secure threaded connection
Specifications
Autoclavable Not Autoclavable
Material Nylon
For Use With 3/8 in. NPT thread
Min. Operating Temperature (Metric) -40°C
Packaging Quantity 6 Pack
Sterilization Method Ethylene Oxide
Thread Size 3/8 in. NPT
Maximum Pressure 200 psi
Max. Operating Temperature (Metric) 93°C
Product Type Threaded Nipple NPT(M) Connector