Learn More
Masterflex™ Nylon Male Pipe Adapter, pack of 100
Product Code. 11760119
Change view
Click to view available options
Thread Size:
1/4 in. NPT(M)
1/8 in. NPT(M)
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size unitSize
11760119 1/4 in. NPT(M) Pack of 100
11740109 1/8 in. NPT(M) Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Product Code. 11760119 missing translation for 'mfr' Masterflex™ missing translation for 'supplierNo' 3062547
Only null left
Add to basket
Add to basket
Designed for quick and easy assembly without tools
  • Adapts from a NPT male thread to a hose barb
  • Barbed fittings are designed for quick and easy assembly without tools
  • Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
  • Available in packs of ten
Specifications
Description Barb x NPT(M) Connection
Autoclavable Not Autoclavable
Material Nylon
Tubing Size 1/4 in.
Hose Barb 1/4 in.
Outlet Port 1/4 in. NPT(M)
Port Size 1/4 in.
Sterilization Method Ethylene Oxide
Thread Size 1/4 in. NPT(M)
Color Translucent
Product Type Male Pipe Adapter