Learn More
Masterflex™ Nylon Male Pipe Adapter
Product Code. 11780139
Click to view available options
Tubing Size:
1/4 in.
1/8 in.
3/8 in.
Unit Size:
Pack of 10
Product Code. 11780139
Brand: Masterflex™ 3062144
Only null left
Add to basket
Add to basket
Designed for quick and easy assembly without tools
  • Adapts from a NPT male thread to a hose barb
  • Barbed fittings are designed for quick and easy assembly without tools
  • Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
  • Available in packs of ten
Specifications
Description Barb x NPT(M) Connection
Autoclavable Not Autoclavable
Material Nylon
Tubing Size 1/8 in.
Hose Barb 1/8 in.
Outlet Port 1/4 in. NPT(M)
Port Size 1/8 in.
Sterilization Method Ethylene Oxide
Thread Size 1/4 in. NPT(M)
Color Translucent
Product Type Male Pipe Adapter