Learn More
Masterflex™ Nylon Male Luer Adapter
Product Code. 15137204
Click to view available options
For Use With:
Male Luer to 1/8-27 Thread
Unit Size:
Pack of 25
Product Code. 15137204
Brand: Masterflex™ 4550586
Only null left
Add to basket
Add to basket
Specifications
Material Nylon
For Use With Male Luer to 1/8-27 Thread
Packaging Quantity 25 Pack
Product Type Male Luer Fitting