Learn More
KD Scientific Nylon Luer Connector Kit
Click to view available options
Material:
Nylon
Unit Size:
Pack of 10
Description
Fittings include:
Luer to Barb, Male Luer Lock (MLL) and Female Luer Lock (FLL) to barbed connector (barb sizes: 1/16 in., 3/32 in., 1/8 in., 5/32 in., 3/16 in., and 1/4 in. ID).
Luer to Luer Connectors: MLT (Male Luer Taper) to MLT, RMLL (Rotating Male Luer Lock) to RMLL, FLL to FLL, FLL to FLL elbow, 3 x FLL T connector, FLL to MLT and both MLL and FLL caps.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Nylon |
| Sterility | Non-sterile |
| Product Type | Connector Kit |