Learn More
Masterflex™ Nylon Female Luer Fitting
Description
- Standard hose barb for use with flexible tubing
- Smooth bore hose barb to reduce flow turbulence and risk of line contamination
- Wide-bore hose barb for enhanced fluid path flow
Specifications
Specifications
| Material | Nylon |
| For Use With | Female Luer x 1/8-27 NPT Elbow |
| Packaging Quantity | 25 Pack |
| Product Type | Female Luer Fitting |