missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Nylon Clamps for Sanitary Fittings
Click to view available options
Width (Metric):
12.7 to 19.05 mm
25.4 to 38.1 mm
Unit Size:
Each
Description
Maintain sanitary conditions with these clamps. Cole-Parmer™ Nylon Clamp for Sanitary Fittings feature excellent chemical resistance and low leachable characteristics.
- Low leachable characteristics
- Sterilize with Ethylene Oxide (ETO)
- Compatible with Tri-Clamp™ fittings
Specifications
Specifications
| Material | Nylon |
| Packaging Quantity | 1 Ea. |
| Width (Metric) | 25.4 to 38.1 mm |
| Product Type | Tubing Clamp |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion