missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Nylon Cable Ties

Product Code. 11835302 Shop All Cole-Parmer Products
Change view
Click to view available options
Color:
Red
White
Length (Metric):
10 cm
14 cm
19 cm
20 cm
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Length (Metric) unitSize
11835302 White 19 cm Pack of 1000
11769549 White 20 cm Pack of 1000
11772753 White 14 cm Pack of 1000
11795748 Red 20 cm Pack of 1000
11722693 White 10 cm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11835302 Supplier Cole-Parmer™ Supplier No. 0683056
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ties continue to hold tension even in oily spray or wet environments. Cole-Parmer™ Nylon Cable Ties feature a durable locking mechanism that holds securely even when used to bundle vibrating or flexing materials.

  • These ties are injection-molded of 6/6 nylon, won’t corrode or stretch
  • Easily apply ties by hand or use a tensioning tool (tool sold separately)
  • Ties are UL-component recognized, and are self-extinguishing as per UL 94 Flam
  • Class: V-2 (models 06830-85 and -95 have a UL flame rating of UL94-HB)
  • All solid design cable ties (A, and E meet military specification MS3367 except the extra-heavy duty models (06830-42 and -99)
  • Ties withstand temperatures to 85°C (185°F) and have a melting point of 257°C (495°F); except for barbed design cable ties which withstand temperatures to 75°C (167°F) and have a melting point of 218°C (425°F)

Specifications

Type Cable Tie
Material Nylon
Color White
Length (Metric) 19 cm
Quantity 1000/Pk.
Certifications/Compliance UL-component recognized
For Use With (Application) Easily apply ties by hand or use a tensioning tool
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.