missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Nylon Biopsy Bags

Ensure unrestricted fluid movement around tissue with Epredia™ Nylon Biopsy Bags.

Brand:  Epredia™ 6774013

Code : 50

Additional Details : Weight : 0.11340kg

 View more versions of this product

Product Code. 12056699

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Reduce risk of small specimen loss
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Remove specimen rapidly with convenient peel-open bag

Dimensions:

  • Small Biopsy Bags measure 1.2 × 1.7″ (31 × 43mm)
  • Medium Biopsy Bags measure 1.8 × 2.6″ (45 × 65mm)
  • Large Biopsy Bags measure 3 × 3.5″ (75 × 85mm)
Specifications

Specifications

Biopsy Bag
Blue
65 mm
2.6 in.
500/Pk.
Nylon
45 mm
1.8 in.
Medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers