missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Nylon Barbed Tee Connector
Click to view available options
Tubing Size:
1 in.
3/4 in.
Unit Size:
Pack of 10
Description
- Ideal for low-pressure applications
- Chemical resistant
- Available in a variety of sizes
- Economical barbed fittings easily connect fluid paths without the use of tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Specifications
| Description | Barb Connections |
| Autoclavable | Not Autoclavable |
| Material | Nylon |
| Overall Length (English) | 3 5/16 in. |
| Tubing Size | 3/4 in. |
| Hose Barb | 3/4 in. |
| Outlet Port | 3/4 in. |
| Port Size | 3/4 in. |
| Sterilization Method | Ethylene Oxide |
| Width (English) | 2 1/32 in. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction