Learn More
Masterflex™ Nylon Barbed Tee Connector
Product Code. 15324188
Click to view available options
Tubing Size:
1 in.
3/4 in.
Unit Size:
Pack of 10
Product Code. 15324188
Brand: Masterflex™ 3062361
Only null left
Add to basket
Add to basket
Designed for quick and easy assembly without tools
  • Ideal for low-pressure applications
  • Chemical resistant
  • Available in a variety of sizes
  • Economical barbed fittings easily connect fluid paths without the use of tools
  • Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Description Barb Connections
Autoclavable Not Autoclavable
Material Nylon
Overall Length (English) 3 5/16 in.
Tubing Size 3/4 in.
Hose Barb 3/4 in.
Outlet Port 3/4 in.
Port Size 3/4 in.
Sterilization Method Ethylene Oxide
Width (English) 2 1/32 in.
Shape T-shape
Color Translucent
Product Type Barbed Tee Connector