missing translation for 'onlineSavingsMsg'
Learn More

NXT629, MedChemExpress

Product Code. p-200096765
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16477547

Brand: Medchem Express HY114263

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NXT629 is a potent, selective, and competitive PPAR-α antagonist, with an IC50 of 77 nM for human PPARα, shows high selectivity over other nuclear hormone receptor, such as PPARδ, PPARγ, ERβ, GR and TRβ, IC50s are 6.0, 15, 15.2, 32.5 and >100 μM, respectively. NXT629 has potent anti-tumor activity and inhibits experimental metastasis of cancer cell in animal models.

NXT629 is a potent, selective, and competitive PPAR-α antagonist, with an IC50 of 77 nM for human PPARα, shows high selectivity over other nuclear hormone receptor, such as PPARδ, PPARγ, ERβ, GR and TRβ, IC50s are 6.0, 15, 15.2, 32.5 and >100 μM, respectively. NXT629 has potent anti-tumor activity and inhibits experimental metastasis of cancer cell in animal models.

Specifications

Chemical Name or Material NXT629
CAS 1454925-59-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C35H39N5O3S
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (204.99 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1)(NC2=CC=C(C3=CC=C(CCCC(N4CC)=NN(CC5=CC=C(C(C)(C)C)C=C5)C4=O)C=C3)N=C2)=O
Molecular Weight (g/mol) 609.78
Formula Weight 609.78
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.