missing translation for 'onlineSavingsMsg'
Learn More

NXT629, MedChemExpress

Product Code. 16477547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477547 5 mg 5mg
16471067 50 mg 50mg
16415606 100 mg 100mg
16414808 10 mg 10mg
16403256 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16477547 Supplier Medchem Express Supplier No. HY114263
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NXT629 is a potent, selective, and competitive PPAR-α antagonist, with an IC50 of 77 nM for human PPARα, shows high selectivity over other nuclear hormone receptor, such as PPARδ, PPARγ, ERβ, GR and TRβ, IC50s are 6.0, 15, 15.2, 32.5 and >100 μM, respectively. NXT629 has potent anti-tumor activity and inhibits experimental metastasis of cancer cell in animal models.

NXT629 is a potent, selective, and competitive PPAR-α antagonist, with an IC50 of 77 nM for human PPARα, shows high selectivity over other nuclear hormone receptor, such as PPARδ, PPARγ, ERβ, GR and TRβ, IC50s are 6.0, 15, 15.2, 32.5 and >100 μM, respectively. NXT629 has potent anti-tumor activity and inhibits experimental metastasis of cancer cell in animal models.

Specifications

Chemical Name or Material NXT629
CAS 1454925-59-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C35H39N5O3S
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (204.99 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1)(NC2=CC=C(C3=CC=C(CCCC(N4CC)=NN(CC5=CC=C(C(C)(C)C)C=C5)C4=O)C=C3)N=C2)=O
Molecular Weight (g/mol) 609.78
Formula Weight 609.78
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.