missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NXT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567222
This item is not returnable.
View return policy
Description
Immunogen sequence: KYRSHWSQGDR EGYQRRSNYY EGPHTSHSSP ADRTREEVVT PTLPEHTATR SQMATSLD Highest antigen sequence identity to the following orthologs - mouse 34%, rat 34%.
Regulator of protein export for NES-containing proteins. Also plays a role in mRNA nuclear export.
Specifications
| NXT2 | |
| Polyclonal | |
| Unconjugated | |
| NXT2 | |
| 6330587F24Rik; AI787442; BM-025; DC9; NTF2-related export protein 2; nuclear transport factor 2 like export factor 2; nuclear transport factor 2-like export factor 2; NXT2; P15-2; protein p15-2; RGD1560204 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55916 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NPJ8 | |
| NXT2 | |
| Recombinant Human NXT2. Recombinant protein control fragment (Product #RP-107858). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction