missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NXPH3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148960
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
May be signaling molecules that resemble neuropeptides. Ligand for alpha-neurexins. Target protein is a member of a family of neuropeptide-like glycoproteins.
Specifications
| NXPH3 | |
| Polyclonal | |
| Unconjugated | |
| NXPH3 | |
| KIAA1159; neurexophilin 3; neurexophilin-3; NPH3; Nxph3; UNQ687/PRO1327 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 104079, 11248, 59315 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O95157, Q91VX5, Q9Z2N5 | |
| NXPH3 | |
| Mouse myeloma cell line NS0-derived recombinant rat Neurexophilin-3/NXPH-3, Gln23-Gly252. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction