missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NXPH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557047
This item is not returnable.
View return policy
Description
Immunogen sequence: LFCDSKEVVH ATEGLDWEDK DAPGTLVGNV VHSRIISPLR LFVKQSPVPK PGPMAYADSM ENFWDWLANI TEIQEPLART KR Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 68%.
Neurexophilin-1 (also known as NPH1 or NXPH1), Neurexophilin-2 (also known as NPH2 or NXPH2) and Neurexophilin-3 (also known as NPH3 or NXPH3) are members of the Neurexophilin family (Neurexophilin-1-4) of neuropeptide-like glycoproteins that are proteolytically processed after synthesis. Neurexophilin-1-3 are secreted proteins that are thought to function as signaling molecules which specifically bind to target proteins, such as neurexin I-alpha (a protein that promotes adhesion between dendrites and axons), and are essential for proper neurotransmitter release. While Neurexophilin-1 is located primarily in spleen tissue, Neurexophilin-2 is expressed primarily in kidney and both Neurexophilin-2 and Neurexophilin-3 are highly expressed in brain. Defects in the gene encoding Neurexophilin-1 may be associated with schizophrenia, a mental disorder characterized by an abnormal perception of reality.
Specifications
| NXPH2 | |
| Polyclonal | |
| Unconjugated | |
| NXPH2 | |
| neurexophilin 2; Neurexophilin2; neurexophilin-2; NPH2; Nxph2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11249 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95156 | |
| NXPH2 | |
| Recombinant protein corresponding to Human NXPH2. Recombinant protein control fragment (Product #RP-96122). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto