missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NXPE3 Monoclonal Antibody (4-E11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA532894
This item is not returnable.
View return policy
Description
NXPE3 Monoclonal Antibody for Western Blot, ICC/IF
Specifications
| NXPE3 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q969Y0 | |
| NXPE3 | |
| Recombinant protein with Human FAM55C aa 155-429. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry, Western Blot | |
| 4-E11 | |
| Unconjugated | |
| NXPE3 | |
| FAM55C; family with sequence similarity 55, member C; MST115; MSTP115; neurexophilin and PC-esterase domain family member 3; neurexophilin and PC-esterase domain family, member 3; NXPE family member 3; NXPE3; Protein FAM55C; UNQ5791/PRO19596 | |
| Mouse | |
| Protein A | |
| RUO | |
| 91775 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction