missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NXPE2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558912
This item is not returnable.
View return policy
Description
Immunogen sequence: KDHTKFSFNL ENHIILNQGN IFKKYSHSET PLCPAVSPKE TELRIKDIME KLDQQ Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 45%.
Specifications
| NXPE2 | |
| Polyclonal | |
| Unconjugated | |
| NXPE2 | |
| FAM55B; family with sequence similarity 55, member B; neurexophilin and PC-esterase domain family member 2; neurexophilin and PC-esterase domain family, member 2; NXPE family member 2; NXPE2; Protein FAM55B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 120406 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96DL1 | |
| NXPE2 | |
| Recombinant protein corresponding to Human NXPE2. Recombinant protein control fragment (Product #RP-97549). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction