missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NXNL2 Antibody (OTI2B11), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73134AF405
This item is not returnable.
View return policy
Description
NXNL2 Monoclonal specifically detects NXNL2 in Human, Mouse, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| NXNL2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NXNL2 | |
| Full length recombinant protein of human NXNL2 (NP_660326) produced in HEK293T cells. | |
| 0.1 mL | |
| 158046.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2B11 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| C9orf121, chromosome 9 open reading frame 121, nucleoredoxin-like 2, nucleoredoxin-like protein 2, RDCVF2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Monkey, Primate | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction