missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NXF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569110
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 86%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 86%.
Could be involved in the export of mRNA from the nucleus to the cytoplasm. Could also have a role in polarized cytoplasmic transport and localization of mRNA in neurons.
Specifications
| NXF5 | |
| Polyclonal | |
| Unconjugated | |
| NXF5 | |
| Nuclear RNA export factor 5; NXF5; TAPL1; TAPL-1; TAP-like protein 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 55998 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H1B4 | |
| NXF5 | |
| synthetic peptide directed towards the middle region of human NXF5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction