missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NXF3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15709790 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15709790 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15709790 Supplier Invitrogen™ Supplier No. PA551832

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: NPAGIPHFVH RELKSEKVEQ IKLAMNQQCD VSQEALDIQR LPFYPDMVNR DTKMASNPRK CMAASLDVHE ENIPTVMSAG EMDKWKGIEP GEKCADRSPV CTTFSDTSSN INSILELFPK LLCLDGQQSP RATLCGTEAH KRL Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 41%.

This gene is one member of a family of nuclear RNA export factor genes. Common domain features of this family are a noncanonical RNP-type RNA-binding domain (RBD), 4 leucine-rich repeats (LRRs), a nuclear transport factor 2 (NTF2)-like domain that allows heterodimerization with NTF2-related export protein-1 (NXT1), and a ubiquitin-associated domain that mediates interactions with nucleoporins. The LRRs and NTF2-like domains are required for export activity. Alternative splicing seems to be a common mechanism in this gene family. The encoded protein of this gene has shortened LRR and ubiquitin-associated domains and its RDB is unable to bind RNA. It is located in the nucleoplasm but is not associated with either the nuclear envelope or the nucleolus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NXF3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NXF3
Gene Accession No. Q9H4D5
Gene Alias Nuclear RNA export factor 3; NXF3; RGD1564923; TAPL3; TAPL-3; TAP-like protein 3
Gene Symbols NXF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NXF3. Recombinant protein control fragment (Product #RP-92215).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56000
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.