missing translation for 'onlineSavingsMsg'
Learn More

NXF1 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18408967 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408967 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408967 Supplier Novus Biologicals Supplier No. NBP222267C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NXF1 Polyclonal antibody specifically detects NXF1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NXF1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp667O0311, Mex67, mRNA export factor TAP, nuclear RNA export factor 1, TAPMEX67, tip associating protein, Tip-associated protein, Tip-associating protein
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 569 and 619 of human Nuclear RNA Export Factor 1 using the numbering given in entry NP_006353.2 (GeneID 10482).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10482
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.