missing translation for 'onlineSavingsMsg'
Learn More

Palifosfamide, MedChemExpress

Product Code. 16416178
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16416178

Brand: Medchem Express HY14798

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Palifosfamide is a novel DNA alkylator and the active metabolite of ifosfamide, with antitumor activity.

Palifosfamide is a novel DNA alkylator and the active metabolite of ifosfamide, with antitumor activity.

Specifications

Chemical Name or Material Palifosfamide
Physical Form Solid
CAS 31645-39-3
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C4H11Cl2N2O2P
For Use With (Application) Cancer-programmed cell death
Synonym Isophosphoramide mustard IPM ZIO-201
Solubility Information DMSO : 40 mg/mL (180.98 mM; Need ultrasonic) ∣H2O : 14.29 mg/mL (64.65 mM; Need ultrasonic)
SMILES O=P(NCCCl)(NCCCl)O
Molecular Weight (g/mol) 221.02
Formula Weight 221.02
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.