missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NVS PAK1 1

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
NVS PAK1 1 is a potent and selective PAK1 inhibitor (IC50 = 5 nM) that displays selective for PAK1 over PAK2 and a panel of 442 kinases. NVS PAK1 1 inhibits autophosphorylation of PAK1 in KRAS-mutated pancreatic cancer cell lines.
A negative control for NVS PAK1 1 is also available; NVS PAK1 C .
Specifications
Specifications
| Chemical Name or Material | (3S)-3-(2-Chloro-5-(2,2-difluoroethyl)-8-fluoro-5H-dibenzo[b,e][1,4]diazepin-11-ylamino)-N-isopropylpyrrolidine-1-carboxamide |
| CAS | 1783816-74-9 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C23H25ClF3N5O |
| Formula Weight | Observed MW: 479.93 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction