missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NVS MLLT-1
Description
NVS-MLLT-1 is a selective inhibitor of YEATS domain of MLLT1 and MLLT3 (IC50 values are 150 and 254 nM for MLLT1 and MLLT3, respectively). It shows selectivity for YEATS2/4 domains and over all bromodomains (CERC2 IC50 > 40 μM).
Specifications
Specifications
| Chemical Name or Material | N-[2-[[(2 S)-2-Methyl-1-pyrrolidinyl]methyl]-1 H-benzimidazol-6-yl]-6-isoquinolinecarboxamide |
| CAS | 2561471-13-2 |
| Quantity | 10 mg |
| Target | Bromodomain Inhibitors |
| Molecular Formula | C23H23N5O |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?