missing translation for 'onlineSavingsMsg'
Learn More

NVP-TNKS656, MedChemExpress

Product Code. 16403406
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16403406

Brand: Medchem Express HY13990

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NVP-TNKS656 is a highly potent, selective, and orally active TNKS2 inhibitor with IC50 of 6 nM, and is > 300 fold selectivity against PARP1 and PARP2.

NVP-TNKS656 is a highly potent, selective, and orally active TNKS2 inhibitor with IC50 of 6 nM, and is > 300 fold selectivity against PARP1 and PARP2.

Specifications

Chemical Name or Material NVP-TNKS656
CAS 1419949-20-4
Color White
Purity Grade Notes Research
Molecular Formula C27H34N4O5
Quantity 25 mg
Synonym TNKS656
Solubility Information DMSO : ≥ 40 mg/mL (80.88 mM)
SMILES O=C(N(CC1CC1)CC2=NC3=C(COCC3)C(N2)=O)CN4CCC(C(C5=CC=C(OC)C=C5)=O)CC4
Molecular Weight (g/mol) 494.58
Formula Weight 494.58
Percent Purity 98.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.