missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NVP CXCR2 20
Potent and selective CXCR2 antagonist
Specifications
| Chemical Name or Material | 4-Cyclopropyl-2-[[(2,3-difluorophenyl)methyl]thio]-1,6-dihydro-6-oxo5-pyrimidinecarbonitrile |
|---|---|
| CAS | 1029521-30-9 |
Description
Potent and selective CXCR2 antagonist (IC50 = 40 nM). Exhibits selectivity for CXCR2 over a panel of 49 other GPCRs. Orally bioavailable.Specifications
| 4-Cyclopropyl-2-[[(2,3-difluorophenyl)methyl]thio]-1,6-dihydro-6-oxo5-pyrimidinecarbonitrile | |
| 1029521-30-9 |
Safety and Handling
Recommended Storage : Store at 4°C
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.