missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NVP-CGM097 sulfate, MedChemExpress
Description
NVP-CGM097 sulfate is a potent and selective MDM2 inhibitor with IC50 of 1.7±0.1 nM for hMDM2.
Specifications
Specifications
| Chemical Name or Material | NVP-CGM097 sulfate |
| CAS | 1313367-56-4 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C38H49ClN4O8S |
| Quantity | 100 mg |
| Synonym | CGM097 sulfate |
| Solubility Information | DMSO : 100 mg/mL (132.04 mM; Need ultrasonic) ∣H2O : 100 mg/mL (132.04 mM; Need ultrasonic) |
| SMILES | ClC(C=C1)=CC=C1[C@@H](C2=CC(OC(C)C)=C(OC)C=C2C3)N(C4=CC=C(N(C[C@@H]5CC[C@@H](N6CC(N(C)CC6)=O)CC5)C)C=C4)C3=O.O=S(O)(O)=O |
| Molecular Weight (g/mol) | 757.34 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?