missing translation for 'onlineSavingsMsg'
Learn More

NVP-CGM097 (stereoisomer), MedChemExpress

Product Code. 16460266
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460266 2 mg 2mg
16419477 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460266 Supplier Medchem Express Supplier No. HY15954A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NVP-CGM097 (stereoisomer) is a stereoisomer of NVP-CGM097, with no special bioactivity. NVP-CGM097 is a potent and selective MDM2 inhibitor.

NVP-CGM097 (stereoisomer) is a stereoisomer of NVP-CGM097, with no special bioactivity. NVP-CGM097 is a potent and selective MDM2 inhibitor.

Specifications

Chemical Name or Material NVP-CGM097 (stereoisomer)
CAS 2070009-54-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C38H47ClN4O4
Quantity 2 mg
Synonym CGM097 stereoisomer (R)-Nvp-Cgm097
Solubility Information DMSO : ≥ 241.5 mg/mL (366.32 mM)
SMILES O=C1N(C2=CC=C(N(C)C[C@H]3CC[C@H](N4CC(N(C)CC4)=O)CC3)C=C2)[C@H](C5=CC=C(Cl)C=C5)C6=C(C=C(OC)C(OC(C)C)=C6)C1
Molecular Weight (g/mol) 659.26
Formula Weight 659.26
Percent Purity 95.83%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.