missing translation for 'onlineSavingsMsg'
Learn More

NVP-BSK805 dihydrochloride, MedChemExpress

Product Code. 16486526
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486526

Brand: Medchem Express HY14722A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NVP-BSK805 dihydrochloride is an ATP-competitive JAK2 inhibitor, with IC50s of 0.48 nM, 31.63 nM, 18.68 nM, and 10.76 nM for JAK2 JH1 (JAK homology 1), JAK1 JH1, JAK3 JH1, and TYK2 JH1, respectively.

NVP-BSK805 dihydrochloride is an ATP-competitive JAK2 inhibitor, with IC50s of 0.48 nM, 31.63 nM, 18.68 nM, and 10.76 nM for JAK2 JH1 (JAK homology 1), JAK1 JH1, JAK3 JH1, and TYK2 JH1, respectively.

Specifications

Chemical Name or Material NVP-BSK805 dihydrochloride
CAS 1942919-79-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H30Cl2F2N6O
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (88.74 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES FC1=C(CN2CCOCC2)C(F)=CC(C3=C4N=C(C5=CN(C6CCNCC6)N=C5)C=NC4=CC=C3)=C1.Cl.Cl
Molecular Weight (g/mol) 563.47
Formula Weight 563.47
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.