missing translation for 'onlineSavingsMsg'
Learn More

NVP-AEW541, MedChemExpress

Product Code. 16497876
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497876 100 mg 100mg
16480578 5 mg 5mg
16478118 10 mg 10mg
16454037 50 mg 50mg
16430356 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497876 Supplier Medchem Express Supplier No. HY50866
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress NVP-AEW541 (AEW541) is a potent inhibitor of IGF-1R with IC50 of 0.15 μM, also inhibits InsR, with IC50 of 0.14 μM.

NVP-AEW541 (AEW541) is a potent inhibitor of IGF-1R with IC50 of 0.15 μM, also inhibits InsR, with IC50 of 0.14 μM.

Specifications

Chemical Name or Material NVP-AEW541
CAS 475489-16-8
Color White
Purity Grade Notes Research
Molecular Formula C27H29N5O
Quantity 100 mg
Synonym AEW541
Solubility Information DMSO : 50 mg/mL (113.75 mM; Need ultrasonic)
SMILES NC1=C2C(N([C@@H]3C[C@@H](C3)CN4CCC4)C=C2C5=CC=CC(OCC6=CC=CC=C6)=C5)=NC=N1
Molecular Weight (g/mol) 439.55
Formula Weight 439.55
Percent Purity 98.45%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.