missing translation for 'onlineSavingsMsg'
Learn More

NVP-ACC789, MedChemExpress

Product Code. 16463777
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463777 50 mg 50mg
16460288 5 mg 5mg
16433666 25 mg 25mg
16417828 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16463777 Supplier Medchem Express Supplier No. HY19624

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NVP-ACC789 is an inhibitor of human VEGFR-1, VEGFR-2 (mouse VEGFR-2), VEGFR-3 and PDGFR-β with IC50s of 0.38, 0.02 (0.23), 0.18, 1.4 μM, respectively.

NVP-ACC789 is an inhibitor of human VEGFR-1, VEGFR-2 (mouse VEGFR-2), VEGFR-3 and PDGFR-β with IC50s of 0.38, 0.02 (0.23), 0.18, 1.4 μM, respectively.

Specifications

Chemical Name or Material NVP-ACC789
CAS 300842-64-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H17BrN4
Quantity 50 mg
Synonym ACC-789 ZK202650
Solubility Information DMSO : 25 mg/mL (61.68 mM; Need ultrasonic)
SMILES CC1=CC=C(NC2=NN=C(CC3=CC=NC=C3)C4=C2C=CC=C4)C=C1Br
Molecular Weight (g/mol) 405.29
Formula Weight 405.29
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.