missing translation for 'onlineSavingsMsg'
Learn More

Rocaglamide, MedChemExpress

Product Code. 16467748
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16467748

Brand: Medchem Express HY19356

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rocaglamide (Roc-A) is isolated from the genus Aglaia and can be used for coughs, injuries, asthma and inflammatory skin diseases.

Rocaglamide is a potent inhibitor of NF-κB activation in T-cells. Rocaglamide is a potent and selective heat shock factor 1 (HSF1) activation inhibitor with an IC50 of ∼50 nM. Rocaglamide inhibits the function of the translation initiation factor eIF4A. Rocaglamide also has anticancer properties in leukemia.

Specifications

Chemical Name or Material Rocaglamide
CAS 84573-16-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H31NO7
Quantity 10 mg
Synonym Roc-A
Solubility Information DMSO : 100 mg/mL (197.80 mM; Need ultrasonic)
SMILES O[C@]12[C@](OC3=CC(OC)=CC(OC)=C32)(C4=CC=C(C=C4)OC)[C@H](C5=CC=CC=C5)[C@H]([C@H]1O)C(N(C)C)=O
Molecular Weight (g/mol) 505.56
Formula Weight 505.56
Percent Purity 96.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.