missing translation for 'onlineSavingsMsg'
Learn More

Nuvenzepine, MedChemExpress

Product Code. 16483159
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483159 1 mg 1mg
16442378 5 mg 5mg
16440179 10 mg 10mg
16430696 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483159 Supplier Medchem Express Supplier No. HYU00119
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Nuvenzepine is an mAChR antagonist, has the potential for gastrospasm treatment.

Nuvenzepine is an mAChR antagonist, has the potential for gastrospasm treatment.

Specifications

Chemical Name or Material Nuvenzepine
CAS 96487-37-5
Color White
Purity Grade Notes Research
Molecular Formula C19H20N4O2
Quantity 1 mg
SMILES O=C1C2=CC=CN=C2N(C(C3CCN(C)CC3)=O)C4=CC=CC=C4N1
Molecular Weight (g/mol) 336.39
Formula Weight 336.39
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.