missing translation for 'onlineSavingsMsg'
Learn More

Melford Nutrient Agar

Product Code. 30018559
Click to view available options
Quantity:
500 g
1 kg
2 kg
Unit Size:
1kg
2kg
500g
This item is not returnable. View return policy

Product Code. 30018559

Brand: Melford N14100500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cultivating a variety of non-fastidious organisms.

For cultivating a variety of non-fastidious organisms.

Dissolve 23g in one liter distilled water. Heat solution to boiling with continuous stirring for one minute. Sterilize by autoclaving at 121°C for 15 minutes. Mix medium with gentle agitation to avoid bubbles prior to pouring plates. Store at room temperature.

Ingredients:

Beef Extract: 3 g/l

Pancreatic Digest of Gelatin: 5 g/l

Agar: 15 g/l

Specifications

Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.