missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUTM2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571090
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
NUTM2A is a protein coding gene.
Specifications
| NUTM2A | |
| Polyclonal | |
| Unconjugated | |
| NUTM2A | |
| FAM22A; family with sequence similarity 22, member A; KIAA2020; NUT family member 2A; NUTM2A; protein FAM22A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 728118 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IVF1 | |
| NUTM2A | |
| synthetic peptide directed towards the C-terminal region of human NUTM2A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction